Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxD3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298721JF646
Description
FoxD3 Polyclonal antibody specifically detects FoxD3 in Human samples. It is validated for Western BlotSpecifications
FoxD3 | |
Polyclonal | |
Western Blot | |
AIS1, forkhead box D3, forkhead box protein D3, HFH2, HNF3/FH transcription factor genesis | |
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human FoxD3. | |
0.1 mL | |
Stem Cell Markers | |
27022 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction