Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585957 Supplier Invitrogen™ Supplier No. PA585957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXD3 Polyclonal Antibody for Western Blot

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. Mutations in this gene cause autoimmune susceptibility 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXD3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene FOXD3
Gene Accession No. Q61060, Q9UJU5
Gene Alias AIS1; CWH3; forkhead box D3; forkhead box protein D3; FOXD3; Genesis; hepatocyte nuclear factor 3 forkhead homolog 2; HFH2; HFH-2; HNF3/FH transcription factor genesis; HNF-3/forkhead homolog 2; VAMAS2
Gene Symbols FOXD3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to of Mouse FOXD3 aa 1-50.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15221, 27022
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.