Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXD4/L2/L3/L4/L5/L6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539172
Description
This gene encodes a member of the forkhead/winged helix-box (FOX) family of transcription factors. FOX transcription factors play critical roles in the regulation of multiple processes including metabolism, cell proliferation and gene expression during ontogenesis. Mutations in this gene are associated with a complex phenotype consisting of dilated cardiomyopathy, obsessive-compulsive disorders, and suicidality. [provided by RefSeq, Mar 2012].
Specifications
FOXD4L | |
Polyclonal | |
Unconjugated | |
FOXD4L6 | |
Forkhead box protein D4-like 2, FOXD4-like 2, Forkhead box protein D4-like 3, FOXD4-like 3, Forkhead box protein D4-like 4, FOXD4-like 4, Forkhead box protein D4-like 5, FOXD4-like 5, Forkhead box protein D4-like 6, FOXD4-like 6, | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2298, 286380, 349334, 653404, 653427 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q12950, Q3SYB3, Q5VV16, Q6VB84, Q8WXT5 | |
FOXD4, FOXD4L3, FOXD4L4, FOXD4L5, FOXD4L6 | |
A synthetic peptide derived from the internal region of human FOXD4D4/L2/L3/L4/L5/L6 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction