Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXD4/L2/L3/L4/L5/L6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101591
Description
Antibody detects endogenous levels of total FOXD4/L2/L3/L4/L5/L6.
This gene encodes a member of the forkhead/winged helix-box (FOX) family of transcription factors. FOX transcription factors play critical roles in the regulation of multiple processes including metabolism, cell proliferation and gene expression during ontogenesis. Mutations in this gene are associated with a complex phenotype consisting of dilated cardiomyopathy, obsessive-compulsive disorders, and suicidality.
Specifications
| FOXD4/L2/L3/L4/L5/L6 | |
| Polyclonal | |
| Unconjugated | |
| FOXD4L6 | |
| bA15J10.2; bA460E7.2; FKHL9; forkhead box D4; forkhead box D4 like 3; forkhead box D4-like 2; forkhead box D4-like 3; forkhead box D4-like 4; forkhead box D4-like 5; forkhead box D4-like 6; forkhead box protein D4; forkhead box protein D4b; Forkhead box protein D4-like 2; Forkhead box protein D4-like 3; Forkhead box protein D4-like 4; forkhead box protein D4-like 5; Forkhead box protein D4-like 6; forkhead, Drosophila, homolog-like 9; forkhead-related activator 5; forkhead-related protein FKHL9; Forkhead-related transcription factor 5; FOXD4; FOXD4A; FOXD4B; FOXD4L2; FOXD4L3; FOXD4L4; FOXD4L5; FOXD4L6; FOXD4-like 2; FOXD4-like 3; FOXD4-like 4; FOXD4-like 5; FOXD4-like 6; FOXD6; FREAC5; FREAC-5; Myeloid factor-alpha; myeloid factor-gamma; winged helix factor-1; winged helix transcription factor beta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2298, 286380, 349334, 653404, 653427 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q12950, Q3SYB3, Q5VV16, Q6VB84, Q8WXT5 | |
| FOXD4, FOXD4L3, FOXD4L4, FOXD4L5, FOXD4L6 | |
| A synthesized peptide derived from human FOXD4(Accession Q12950), corresponding to amino acid residues N176-N226. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction