Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106665 Supplier Invitrogen™ Supplier No. PA5106665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FOXD4.

This gene encodes a member of the forkhead/winged helix-box (FOX) family of transcription factors. FOX transcription factors play critical roles in the regulation of multiple processes including metabolism, cell proliferation and gene expression during ontogenesis. Mutations in this gene are associated with a complex phenotype consisting of dilated cardiomyopathy, obsessive-compulsive disorders, and suicidality.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXD4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FOXD4
Gene Accession No. Q12950, Q60688
Gene Alias Fkh2; Fkhl9; forkhead box D4; forkhead box protein D4; forkhead homolog 2; forkhead, Drosophila, homolog-like 9; forkhead-related activator 5; forkhead-related protein FKHL9; Forkhead-related transcription factor 5; Foxd4; FOXD4A; Freac5; FREAC-5; hepatocyte nuclear factor 3 forkhead homolog 6; HFH-6; myeloid factor-alpha; Transcription factor FKH-2
Gene Symbols FOXD4
Host Species Rabbit
Immunogen A synthesized peptide derived from human FOXD4(Accession Q12950), corresponding to amino acid residues N176-N226.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14237, 2298
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.