Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113634
Description
Immunogen sequence: MNSARAGHLR STPPPSPLSS DQDEVEIDVL AEEEDGDQTE EEDDEEESHK For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Mouse: 100%; Rat: 77%.
This gene encodes a member of the forkhead/winged helix-box (FOX) family of transcription factors. FOX transcription factors play critical roles in the regulation of multiple processes including metabolism, cell proliferation and gene expression during ontogenesis. Mutations in this gene are associated with a complex phenotype consisting of dilated cardiomyopathy, obsessive-compulsive disorders, and suicidality.
Specifications
| FOXD4 | |
| Polyclonal | |
| Unconjugated | |
| FOXD4 | |
| Fkh2; Fkhl9; forkhead box D4; forkhead box protein D4; forkhead homolog 2; forkhead, Drosophila, homolog-like 9; forkhead-related activator 5; forkhead-related protein FKHL9; Forkhead-related transcription factor 5; Foxd4; FOXD4A; Freac5; FREAC-5; hepatocyte nuclear factor 3 forkhead homolog 6; HFH-6; myeloid factor-alpha; Transcription factor FKH-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14237 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q60688 | |
| FOXD4 | |
| Synthetic peptide directed towards the N terminal region of mouse Foxd4 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction