Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXD4L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114463
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114463 Supplier Invitrogen™ Supplier No. PA5114463
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive controls that may be used include: A20 Cell Lysate, MCF7 Cell Slide.

This gene is a member of the forkhead/winged-helix (FOX) family of transcription factors with highly conserved FOX DNA-binding domains. Members of the FOX family of transcription factors are regulators of embryogenesis and may play a role in human cancer. This gene lies in a region of chromosome 2 that surrounds the site where two ancestral chromosomes fused to form human chromosome 2. This region is duplicated elsewhere in the human genome, primarily in subtelomeric and pericentromeric locations, thus mutiple copies of this gene have been found. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXD4L1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FOXD4L1
Gene Accession No. Q9NU39
Gene Alias bA395L14.1; forkhead box D4 like 1; forkhead box D4-like 1; forkhead box protein D4-like 1; FOXD4L1; FOXD4-like 1; FOXD5
Gene Symbols FOXD4L1
Host Species Rabbit
Immunogen FOXD4L1 antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human FOXD4L1. The immunogen is located within the last 50 amino acids of FOXD4L1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200350
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.