Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXE1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001271
Description
FOXE1 Polyclonal specifically detects FOXE1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
FOXE1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FKHL15HFKL5, forkhead box E1 (thyroid transcription factor 2), forkhead box E2, Forkhead box protein E2, forkhead, drosophila, homolog-like 15, Forkhead-related protein FKHL15, HFKH4Thyroid transcription factor 2, TITF2HNF-3/fork head-like protein 5, TTF2forkhead box protein E1, TTF-2FOXE2 | |
Goat | |
Affinity Purified | |
RUO | |
2304 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Immunohistochemistry 2 - 5 ug/ml, Immunohistochemistry-Paraffin 2 - 5 ug/ml, Peptide ELISA Detection limit 1:8000 | |
NP_004464. | |
FOXE1 | |
Peptide with sequence C-AYPGGIDRFVSAM corresponding to C-Terminus according to NP_004464.2. | |
0.1 mg | |
Primary | |
Cross reacts with Human. Expected from sequence similarity: Mouse, Dog, Pig and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction