Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540519 Supplier Invitrogen™ Supplier No. PA540519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PEPPCCAAPD AAAAAFPPCA AAASPPLYSQ VPDRLVLPAT RPGPGPLPAE Sequence homology: Human: 100%.

Forkhead Box Protein E3 (FOXE3, forkhead-related protein FKHL12, forkhead-related transcription factor 8) is a forkhead/winged helix transcription factor, which is expressed in the developing lens from the start of lens placode induction and becomes restricted to the anterior proliferating cells when lens fiber differentiation begins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXE3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FOXE3
Gene Accession No. Q13461
Gene Alias dyl; dysgenetic lens; FKHL12; forkhead box E3; forkhead box protein E3; forkhead, drosophila, homolog-like 12; forkhead-related protein FKHL12; Forkhead-related transcription factor 8; Foxe3; FREAC8; FREAC-8; Hepatocyte nuclear factor 3 forkhead homolog 7; HFH7; HFH-7; HNF3; rct; Rinshoken cataract
Gene Symbols FOXE3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human FOXE3 (aa 235-284).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2301
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.