Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxF1 Antibody [Janelia Fluor™ 525], Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals AF4798JF525
Description
FoxF1 Polyclonal antibody specifically detects FoxF1 in Human samples. It is validated for Western BlotSpecifications
FoxF1 | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Antigen Affinity-purified | |
RUO | |
2294 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 525 | |
FKHL5MGC105125, forkhead box F1, forkhead box protein F1, Forkhead, drosophila, homolog-like 5, Forkhead-related activator 1, Forkhead-related protein FKHL5, Forkhead-related transcription factor 1, FREAC-1, FREAC1ACDMPV | |
E. coli-derived recombinant human FoxF1, Met154-Met379, Accession # Q12946 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction