Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxF1 Antibody (SR1668), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32224150UL
Description
FoxF1 Monoclonal antibody specifically detects FoxF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
FoxF1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
2294 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1668 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
FKHL5MGC105125, forkhead box F1, forkhead box protein F1, Forkhead, drosophila, homolog-like 5, Forkhead-related activator 1, Forkhead-related protein FKHL5, Forkhead-related transcription factor 1, FREAC-1, FREAC1ACDMPV | |
A synthesized peptide derived from human FoxF1 (Uniprot #: Q12946) | |
50 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction