Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583039 Supplier Invitrogen™ Supplier No. PA583039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GFRRKCQALK PMYSMMNGLG FNHLPDTYGF QGSAGGLSCP PNSLALEGGL GMMNGHLPGN VDGMALPSHS VPHLPSNGGH SYMGG Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 94%.

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the regulation of pulmonary genes as well as embryonic development. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Foxf1
Gene Accession No. Q12946
Gene Alias ACDMPV; AI450827; FKHL5; forkhead box F1; forkhead box F1a; forkhead box protein F1; forkhead box protein F1; LOW QUALITY PROTEIN: forkhead box protein F1; Forkhead, drosophila, homolog-like 5; forkhead-related activator 1; Forkhead-related protein FKHL5; forkhead-related transcription factor 1; Foxf1; Foxf1a; FREAC1; Freac-1; Hepatocyte nuclear factor 3 forkhead homolog 8; Hfh8; HFH-8; HNF-3/forkhead homolog 8
Gene Symbols Foxf1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FOXF1. Recombinant protein control fragment (Product #RP-100730).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.