Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXF1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA540516

Catalog No. PIPA540516


Only null left
Add to Cart

Description

Description

Peptide sequence: PCNPAANPLS GSLSTHSLEQ PYLHQNSHNA PAELQGIPRY HSQSPSMCDR Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 92%

FOXF1 Polyclonal antibody specifically detects FOXF1 in Human samples. It is validated for Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOXF1
Polyclonal
Unconjugated
Foxf1
ACDMPV; AI450827; FKHL5; forkhead box F1; forkhead box F1a; forkhead box protein F1; forkhead box protein F1; LOW QUALITY PROTEIN: forkhead box protein F1; Forkhead, drosophila, homolog-like 5; forkhead-related activator 1; Forkhead-related protein FKHL5; forkhead-related transcription factor 1; Foxf1; Foxf1a; FREAC1; Freac-1; Hepatocyte nuclear factor 3 forkhead homolog 8; Hfh8; HFH-8; HNF-3/forkhead homolog 8
Rabbit
Protein A
RUO
2294
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q12946
Foxf1
Synthetic peptide directed towards the C-terminal of human FOXF1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.