Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxF1 Rabbit anti-Human, Mouse, Clone: 8F6G3, Novus Biologicals™
SDP

Catalog No. p-7248128 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168366 20 μg
NB168365 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168366 Supplier Novus Biologicals Supplier No. NBP31663720UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FoxF1 Monoclonal antibody specifically detects FoxF1 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen FoxF1
Applications Western Blot
Classification Monoclonal
Clone 8F6G3
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FKHL5MGC105125, forkhead box F1, forkhead box protein F1, Forkhead, drosophila, homolog-like 5, Forkhead-related activator 1, Forkhead-related protein FKHL5, Forkhead-related transcription factor 1, FREAC-1, FREAC1ACDMPV
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FoxF1 (Q12946). MSSAPEKQQPPHGGGGGGGGGGGAAMDPASSGPSKAKKTNAGIRRPEKPPYSYIALIVMAIQSSPTKRLTLSEIYQFLQSRFPFFRGSYQGWKNSVRHNL
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2294
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.