Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540420
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540420 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540420 Supplier Thermo Scientific Supplier No. PA540420
Add to Cart
Add to Cart

FOXF2 Polyclonal Antibody

FOXF2 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXF2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FOXF2
Gene Accession No. Q12947
Gene Alias Fkh20; FKHL6; forkhead box F2; forkhead box protein F2; forkhead homolog 20; forkhead-like 6; forkhead-related activator 2; Forkhead-related protein FKHL6; forkhead-related transcription factor 2; FOXF2; FREAC2; FREAC-2; LOW QUALITY PROTEIN: forkhead box protein F2; LUN; Protein LUN
Gene Symbols FOXF2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FOXF2 (aa 52-101).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2295
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.