Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXG1 Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody

Supplier:  Novus Biologicals NB1001099

Catalog No. NB1001099


Only null left
Add to Cart

Description

Description

FOXG1 Polyclonal specifically detects FOXG1 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

FOXG1A/FOXG1B
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
FOXG1
Peptide with sequence C-HQNQGSSSNPLIH corresponding to C-Terminus according to NP_005240.3.
0.1 mg
Growth and Development, Neuronal Cell Markers
2290
Human, Mouse
IgG
Western Blot, ELISA
0.5 mg/mL
Western Blot 0.3 - 10 ug/ml, Peptide ELISA Detection limit 1:64000
BF-1, BF1HBF-2, BF2, BF-2, Brain factor 1, Brain factor 2, FHKL3, FKHL1forkhead-like 1, FKHL2, FKHL3, FKHL4, forkhead box G1, forkhead box G1A, forkhead box G1B, forkhead box G1C, Forkhead box protein G1A, Forkhead box protein G1B, Forkhead box protein G1C, forkhead-like 3, forkhead-like 4, Forkhead-related protein FKHL1, Forkhead-related protein FKHL2, Forkhead-related protein FKHL3, FOXG1A, FOXG1Bforkhead-like 2, FOXG1C, HBF-1, hBF-2, HBF-3, HFK1HBF-G2, HFK2HBF2, HFK3KHL2, oncogene QIN, QINFKH2forkhead box protein G1
Goat
Affinity Purified
RUO
Primary
This will recognize both human FOXG1A and FOXG1B, which are almost completely identical. Cross reacts with Human and Mouse. Expected from sequence similarity: Rat, Dog and Cow.
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only