Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541493 Supplier Invitrogen™ Supplier No. PA541493
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MLDMGDRKEV KMIPKSSFSI NSLVPEAVQN DNHHASHGHH NSHHPQHHHH Sequence homology: Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 93%.

FOXG1B belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of FOXG1B has not yet been determined; however, it may play a role in the development of the brain and telencephalonThis gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXG1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FOXG1
Gene Accession No. P55316
Gene Alias 2900064B05Rik; Bf1; BF-1; BF1A; BF2; BF-2; brain factor 1; Brain factor 2; FHKL3; FKH2; FKHL1; FKHL2; FKHL3; FKHL4; forkhead box G1; forkhead box G1B; forkhead box protein G1; Forkhead box protein G1A; Forkhead box protein G1B; Forkhead box protein G1C; forkhead-like 1; forkhead-like 2; forkhead-like 3; forkhead-like 4; forkhead-like transcription factor BF-1; forkhead-related protein FKHL1; Forkhead-related protein FKHL2; Forkhead-related protein FKHL3; FOXG1; FOXG1A; FOXG1B; FOXG1C; HBF-1; HBF2; HBF-2; HBF-3; HBF-G2; Hfh9; Hfhbf1; HFK1; HFK2; HFK3; HNF-3/forkhead homolog, brain factor 1; KHL2; oncogene QIN; QIN; RATBF1A; transcription factor BF-1
Gene Symbols FOXG1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human FOXG1B (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2290
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.