Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXG1 Rabbit anti-Human, Polyclonal, Abnova™

Catalog No. 89033373 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-373 Supplier Abnova Corporation Supplier No. PAB30085
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against synthetic peptide of human FOXG1.

  • High specificity
  • High reactivity

Specifications

Antigen FOXG1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of human FOXG1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (1.25 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (2% sucrose, 0.09% sodium azide).
Gene Accession No. NM_005249
Gene Alias BF1/BF2/FHKL3/FKH2/FKHL1/FKHL2/FKHL3/FKHL4/FOXG1A/FOXG1B/FOXG1C/HBF-1/HBF-2/HBF-3/HBF-G2/HBF2/HFK1/HFK2/HFK3/KHL2/QIN
Gene Symbols FOXG1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human FOXG1.
Purification Method Protein A purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2290
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.