Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxH1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | FoxH1 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FoxH1 Polyclonal specifically detects FoxH1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
FoxH1 | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Fast-1, FAST1forkhead box protein H1, Fast-2, FAST2, Forkhead activin signal transducer 1, Forkhead activin signal transducer 2, forkhead activin signal transducer-1, forkhead box H1, hFAST-1, TGF-beta/activin signal transducer | |
FOXH1 | |
IgG | |
Affinity Purified | |
40 kDa |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
PBS with 0.02% Sodium Azide | |
8928 | |
Antibody was raised against a 19 amino acid peptide near the amino terminus of human FOXH1. The immunogen is located within the first 50 amino acids of FOXH1. Amino Acid Squence: CQPPKRRKKRYLRHDKPPYT | |
Primary | |
FOXH1 antibody is predicted to not cross-react with any other members of the forkhead box family. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title