Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534474
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534474 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534474 Supplier Invitrogen™ Supplier No. PA534474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human liver tissue lysate. PA5-34474 can be used with blocking peptide PEP-1516.

FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXH1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene FOXH1
Gene Accession No. O75593, O88621
Gene Alias contains winged-helix domain; etID62177.12; fast; Fast1; Fast-1; FAST1 transcription factor; FAST2; fast-2; fe26b06; Forkhead activin signal transducer 1; forkhead activin signal transducer 2; forkhead activin signal transducer FAST; forkhead activin signal transducer-1; forkhead box H1; forkhead box protein H1; Foxh1; hFAST-1; schmalspur; Schmalspur protein; SUR; TGF-beta/activin signal transducer; transcription factor; uncle freddy; unf; wu:fe26b06; wu:fi30c08; wu:fi32b01
Gene Symbols FOXH1
Host Species Rabbit
Immunogen A 19 amino acid peptide near the amino terminus of human FOXH1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14106, 300054, 8928
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.