Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572644
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572644 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572644 Supplier Invitrogen™ Supplier No. PA572644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXH1 Polyclonal Antibody for Western Blot

FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXH1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FOXH1
Gene Accession No. Q9I9E1
Gene Alias contains winged-helix domain; etID62177.12; fast; Fast1; Fast-1; FAST1 transcription factor; FAST2; fast-2; fe26b06; Forkhead activin signal transducer 1; forkhead activin signal transducer 2; forkhead activin signal transducer FAST; forkhead activin signal transducer-1; forkhead box H1; forkhead box protein H1; Foxh1; hFAST-1; schmalspur; Schmalspur protein; SUR; TGF-beta/activin signal transducer; transcription factor; uncle freddy; unf; wu:fe26b06; wu:fi30c08; wu:fi32b01
Gene Symbols FOXH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 313-334 amino acids from the Central region of DANRE foxh1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57930
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.