Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxH1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | FoxH1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
FoxH1 Polyclonal specifically detects FoxH1 in Human samples. It is validated for Western Blot.Specifications
FoxH1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Stem Cell Markers | |
PBS buffer, 2% sucrose | |
8928 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Fast-1, FAST1forkhead box protein H1, Fast-2, FAST2, Forkhead activin signal transducer 1, Forkhead activin signal transducer 2, forkhead activin signal transducer-1, forkhead box H1, hFAST-1, TGF-beta/activin signal transducer | |
The immunogen is a synthetic peptide directed towards the N terminal region of human FoxH1 (NP_003914). Peptide sequence MGPCSGSRLGPPEAESPSQPPKRRKKRYLRHDKPPYTYLAMIALVIQAAP | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title