Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXI1 Antibody (PCRP-FOXL1-1F8), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314224V
This item is not returnable.
View return policy
Description
FOXI1 Monoclonal antibody specifically detects FOXI1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| FOXI1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2299 | |
| Store at 4°C in the dark. | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PCRP-FOXL1-1F8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| FKHL10HFH3, forkhead box I1, forkhead box protein I1, forkhead-like 10, forkhead-related activator 6, Forkhead-related protein FKHL10, Forkhead-related transcription factor 6, FREAC-6, FREAC6FKH10, Hepatocyte nuclear factor 3 forkhead homolog 3, HFH-3, HNF-3/fork-head homolog 3, HNF-3/fork-head homolog-3, MGC34197 | |
| Recombinant fragment (around aa104-258) of human FOXI1 protein (exact sequence is proprietary) (Uniprot: Q12951) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction