Learn More
Invitrogen™ FOXI1 Monoclonal Antibody (OTI5D3)

Description
FOXI1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | FOXI1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI5D3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FOXI1 |
| Gene Accession No. | Q12951 |
| Gene Alias | 5830401E05Rik; Fkh10; FKHL10; Forkhead; forkhead box I1; forkhead box protein I1; forkhead homolog 10; forkhead-like 10; forkhead-related activator 6; forkhead-related protein FKHL10; Forkhead-related transcription factor 6; Foxi1; FREAC6; FREAC-6; Hepatocyte nuclear factor 3 forkhead homolog 3; HFH3; HFH-3; HNF-3/forkhead homolog 3; HNF-3/fork-head homolog 3; HNF-3/fork-head homolog-3; MGC34197 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.