Learn More
FOXI1 Mouse anti-Human, Clone: OTI1H9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800147

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it is possible that this gene plays an important role in the development of the cochlea and vestibulum, as well as embryogenesis. Mutations in this gene may be associated with the common cavity phenotype. Two transcript variants encoding different isoforms have been found for this gene.Specifications
FOXI1 | |
Monoclonal | |
Unconjugated | |
FOXI1 | |
FKH10, FKHL10, FREAC-6, FREAC6, HFH-3, HFH3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2299 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1H9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q12951 | |
FOXI1 | |
Human recombit protein fragment corresponding to amino acids 111-350 of human FOXI1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.