Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598443
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598443 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598443 Supplier Thermo Scientific Supplier No. PA598443
Add to Cart
Add to Cart

FOXI1 Polyclonal Antibody

FOXI1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXI1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FOXI1
Gene Accession No. Q12951
Gene Alias 5830401E05Rik; Fkh10; FKHL10; Forkhead; forkhead box I1; forkhead box protein I1; forkhead homolog 10; forkhead-like 10; forkhead-related activator 6; forkhead-related protein FKHL10; Forkhead-related transcription factor 6; Foxi1; FREAC6; FREAC-6; Hepatocyte nuclear factor 3 forkhead homolog 3; HFH3; HFH-3; HNF-3/forkhead homolog 3; HNF-3/fork-head homolog 3; HNF-3/fork-head homolog-3; MGC34197
Gene Symbols FOXI1
Host Species Rabbit
Immunogen Recombinant Human Forkhead box protein I1 protein (183-343AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2299
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.