Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568919 Supplier Invitrogen™ Supplier No. PA568919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Rat: 86%.

FOXI2 (Forkhead Box I2) is a Protein Coding gene. Diseases associated with FOXI2 includeBeare-Stevenson Cutis Gyrata SyndromeandCardiofaciocutaneous Syndrome 1. Gene Ontology (GO) annotations related to this gene includeDNA-binding transcription factor activityandDNA-binding transcription factor activity, RNA polymerase II-specific. An important paralog of this gene isFOXI3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXI2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Foxi2
Gene Accession No. Q3I5G5
Gene Alias B130055A05Rik; Fkhl5; forkhead box I2; forkhead box protein I2; Foxf1; Foxi2; hepatocyte nuclear factor 3 forkhead homolog 5; HFH-5; HNF-3/fork-head homolog-5 (HFH-5)
Gene Symbols Foxi2
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse FOXI2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270004
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.