Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxJ1/HFH4 Antibody (407003), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB3619N
Description
FoxJ1/HFH4 Monoclonal antibody specifically detects FoxJ1/HFH4 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
FoxJ1/HFH4 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
407003 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
FKHL13forkhead transcription factor HFH-4, forkhead box J1, forkhead-like 13, Forkhead-related protein FKHL13, Hepatocyte nuclear factor 3 forkhead homolog 4, HFH4fork head homologue 4, HFH-4forkhead box protein J1, MGC35202 | |
E. coli-derived recombinant human FoxJ1, Lys306-Leu421, Accession # Q92949.3 | |
0.1 mL | |
Cancer, Transcription Factors and Regulators | |
2302 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction