Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxJ1/HFH4 Antibody (CL3991), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP25903325UL
Description
FoxJ1/HFH4 Monoclonal specifically detects FoxJ1/HFH4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FoxJ1/HFH4 | |
Monoclonal | |
Immunohistochemistry-Paraffin 1:200 - 1:500 | |
FKHL13forkhead transcription factor HFH-4, forkhead box J1, forkhead-like 13, Forkhead-related protein FKHL13, Hepatocyte nuclear factor 3 forkhead homolog 4, HFH4fork head homologue 4, HFH-4forkhead box protein J1, MGC35202 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
FOXJ1 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PREKDEPGKGGFWRIDPQYAERLLSGAFKKRRLPPVHIHPAFARQAAQEPSAVPRAGPLTVNTEAQQLLREFEEATGEAGWGAGEGRLGHKRKQPLPKRVAKVPRPPSTLLPTPEEQGELEPLKG | |
25 μL | |
Cancer, Transcription Factors and Regulators | |
2302 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction