Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXJ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585557
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585557 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585557 Supplier Invitrogen™ Supplier No. PA585557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (89%), Rat (89%), Bovine (95%). Positive Control: human FOXJ1-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Forkhead-box J1 (FOXJ1) is a 421-amino acid transcription factor that suppresses T cell activity and spontaneous autoimmunity, through the repression of NF B activity. FOXJ1 also inhibits the humoral immune response in B cells. FOXJ1 deficiency in B cells results in spontaneous and accentuated germinal center formation and is implicated in the development of pathogenic autoantibodies and accentuated responses to immunizations. Abnormal expression of FOXJ1 may be associated with autoimmune diseases and/or other inflammatory diseases. FOXJ1 is also required for cilia formation and left-right axis determination because it increases calpastatin expression, a protein necessary for the ability of basal bodies to anchor to the apical cytoskeleton. FOXJ1 expression may function as an early marker of epithelial cell differentiation, recovery, and function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXJ1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene FOXJ1
Gene Accession No. Q92949
Gene Alias FKHL13; FKHL-13; fork head homologue 4; forkhead box J1; forkhead box protein J1; forkhead transcription factor HFH-4; forkhead-like 13; forkhead-related protein FKHL13; Foxj1; Hepatocyte nuclear factor 3 forkhead homolog 4; Hfh4; HFH-4; HNF-3/forkhead homolog 4; MGC35202
Gene Symbols FOXJ1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FOXJ1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2302
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.