Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxJ3 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB260993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260993 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB260993 Supplier Novus Biologicals Supplier No. NBP171861AF750
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FoxJ3 Polyclonal antibody specifically detects FoxJ3 in Human samples. It is validated for Western Blot

Specifications

Antigen FoxJ3
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias forkhead box J3, forkhead box protein J3, KIAA1041, MGC165036, MGC176686
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 25 and 75 of human Forkhead Box J3 using the numbering given in entry NP_055762.3 (GeneID 22887).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22887
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.