Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxJ3 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171861JF646
Description
FoxJ3 Polyclonal antibody specifically detects FoxJ3 in Human samples. It is validated for Western BlotSpecifications
FoxJ3 | |
Polyclonal | |
Western Blot | |
forkhead box J3, forkhead box protein J3, KIAA1041, MGC165036, MGC176686 | |
The immunogen for this product maps to a region between residue 25 and 75 of human Forkhead Box J3 using the numbering given in entry NP_055762.3 (GeneID 22887). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
22887 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction