Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXJ3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549718 Supplier Invitrogen™ Supplier No. PA549718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous level of total FOXJ3 protein.

Transcriptional activator of MEF2C involved in the regulation pf adult muscle fiber type identity and skeletal muscle regeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXJ3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene FOXJ3
Gene Accession No. Q8BUR3, Q9UPW0
Gene Alias C330039G02Rik; Fhd6; forkhead box J3; forkhead box protein J3; forkhead domain gene 6; FOXJ3; Kiaa1041
Gene Symbols FOXJ3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from 610-622 amino acids of Human forkhead box J1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22887, 230700, 313554
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.