Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599303
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599303 Supplier Invitrogen™ Supplier No. PA599303
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FOXL1.

The forkhead domain is a 100-amino acid monomeric DNA binding motif originally identified as a region of homology between the Drosophila forkhead protein and rat HNF3. Pierrou et al. (1994) identified 7 human genes containing forkhead domains and designated them forkhead related activators (FREAC) 1 through 7.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FOXL1
Gene Accession No. Q12952
Gene Alias FKH11; Fkh6; fkh-6; FKHL10; FKHL11; forkhead box L1; forkhead box protein L1; forkhead homolog 6; Forkhead related transcription factor; forkhead-like 11; forkhead-related protein FKHL11; Forkhead-related transcription factor 7; Foxl1; FREAC6; FREAC7; FREAC-7; JFJ-3; Transcription factor FKH-6
Gene Symbols FOXL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FOXL1(Accession Q12952), corresponding to amino acid residues L130-G180.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2300
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.