Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXL2 Antibody, Allophycocyanin/Cy7, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP270013APCCY7
Description
FOXL2 Polyclonal specifically detects FOXL2 in Human samples. It is validated for Flow Cytometry.Specifications
FOXL2 | |
Polyclonal | |
Flow Cytometry | |
BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3 | |
Rabbit | |
Affinity Purified | |
Epigenetics | |
668.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
FOXL2 | |
Synthetic peptide made to a C-terminal portion of the human FOXL2 protein (between amino acids 350-376) [UniProt P58012] | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction