Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXL2 Antibody (FOXL2/8701R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32050320UG
Description
FOXL2 Monoclonal antibody specifically detects FOXL2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FOXL2 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3 | |
Recombinant full-length human FOXL2 protein | |
20 μg | |
Epigenetics | |
668 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
FOXL2/8701R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction