Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXL2 Antibody (SR1880), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32203250UL
Description
FOXL2 Monoclonal antibody specifically detects FOXL2 in Human samples. It is validated for Western BlotSpecifications
FOXL2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1880 | |
Western Blot 1:500-1:2000 | |
BPES, BPES1PINTO, forkhead box L2, forkhead box protein L2, forkhead transcription factor FOXL2, PFRK, POF3 | |
A synthesized peptide derived from human FOXL2 (Uniprot #: P58012) | |
50 μg | |
Epigenetics | |
668 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction