Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXL2 Monoclonal Antibody (CL12753)
GREENER_CHOICE

Catalog No. PIMA546525
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546525 Supplier Invitrogen™ Supplier No. MA546525
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 74%.

FOXL2 is a winged helix/forkhead transcription factor that is highly conserved in human, goat, mouse, and certain aquatic species. Given its cross-species conservation, it appears to play a critical role evolutionarily, likely in ovarian differentiation in mammals and in ovarian somatic cell differentiation and in further follicle development and/or maintenance. Immunohistochemical data shows that FOXL2 is a nuclear protein specifically expressed in eyelids and in fetal and adult ovarian follicular cells. It does not undergo any major posttranslational modifications. FOXL2 is also found to be associated with blepharo-phimosis/ptosis/epicanthus inverses syndrome (BPES). There are two forms of BPES. Type I BPES shows eyelid abnormal-ities that are associated with ovarian failure. In type II BPES, only eyelid defects are found. Studies have shown that FOXL2 is expressed in the mesenchyme of developing mouse eyelids and in adult ovarian follicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXL2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL12753
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FOXL2
Gene Accession No. P58012
Gene Alias AU045128; Blepharophimosis; BPES; BPES1; epicanthus inversus; forkhead box L2; forkhead box protein L2; forkhead transcription factor; forkhead transcription factor FOXL2; forkhead transcription factor L2; Foxl2; LUSTR2; MGC14393; Pfrk; P-Frk; PINTO; pituitary forkhead factor; POF3; putative transcription factor foxl2
Gene Symbols FOXL2
Host Species Mouse
Immunogen Synthetic Peptide.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 668
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.