Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR108 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA132792
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA132792 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA132792 Supplier Invitrogen™ Supplier No. PA132792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-32792 detects GPR108 from human samples.

GPR108 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR108
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR108
Gene Accession No. Q148L1, Q6P6V6, Q91WD0, Q9NPR9
Gene Alias 1810015L19Rik; AA589464; C79132; G protein-coupled receptor 108; GPR108; Lung seven transmembrane receptor 2; LUSTR2; Protein GPR108
Gene Symbols GPR108
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from 2nd cytoplasmic domain of human GPR108.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100764254, 316136, 507830, 56927, 78308
Target Species Human, Mouse, Rat, Bovine, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.