Learn More
FOXL2 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | FOXL2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | FOXL2 |
| Gene Accession No. | P58012 |
| Gene Alias | AU045128; Blepharophimosis; BOS_1244; BPES; BPES1; epicanthus inversus; forkhead box L2; Forkhead box protein L2; forkhead transcription factor FOXL2; forkhead transcription factor L2; Foxl2; LUSTR2; MGC14393; Pfrk; P-Frk; PINTO; Pituitary forkhead factor; POF3; putative transcription factor foxl2 |
| Gene Symbols | FOXL2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.