Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXL2NB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567199
Description
Immunogen sequence: AGPRNTRRLPA PERERIELAA TLCLEGWPLR CLASKGK Highest antigen sequence identity to the following orthologs - mouse 33%, rat 34%.
FOXL2NB is a protein coding gene.
Specifications
FOXL2NB | |
Polyclonal | |
Unconjugated | |
FOXL2NB | |
C3orf72; FLJ43329 protein; FOXL2 neighbor; FOXL2 neighbor protein; FOXL2NB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
401089 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZUU3 | |
FOXL2NB | |
Recombinant Human FOXL2NB. Recombinant protein control fragment (Product #RP-107836). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction