Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXL2NB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566360 Supplier Invitrogen™ Supplier No. PA566360
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PALVKKRMPDA CTLGRAGIGL PKMCLHMAVR HSKAQKTGPG ILQQRQKPPA PRASGGPALL GKRRGCSEA Highest antigen sequence identity to the following orthologs - mouse 30%, rat 29%.

FOXL2NB is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXL2NB
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FOXL2NB
Gene Accession No. Q6ZUU3
Gene Alias C3orf72; FLJ43329 protein; FOXL2 neighbor; FOXL2 neighbor protein; FOXL2NB
Gene Symbols FOXL2NB
Host Species Rabbit
Immunogen Recombinant Human FOXL2NB. Recombinant protein control fragment (Product #RP-105165).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401089
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.