Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxM1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074660FR
Description
FoxM1 Polyclonal antibody specifically detects FoxM1 in Human samples. It is validated for Western BlotSpecifications
FoxM1 | |
Polyclonal | |
Western Blot | |
FKHL16trident, forkhead box M1, forkhead box protein M1, Forkhead, drosophila, homolog-like 16, forkhead-like 16, Forkhead-related protein FKHL16, Hepatocyte nuclear factor 3 forkhead homolog 11, HFH11FOXM1B, HFH-11M-phase phosphoprotein 2, HNF-3, HNF-3/fork-head homolog 11, INS-1, MPHOSPH2, MPP-2, MPP2MPM-2 reactive phosphoprotein 2, PIG29, TGT3, Transcription factor Trident, TRIDENT, WIN, Winged-helix factor from INS-1 cells | |
The immunogen recognized by this antibody maps to a region between residue 713 and 763 of human forkhead box M1 using the numbering given in entry NP_973731.1 (GeneID 2305). | |
0.1 mL | |
Cancer, Mitotic Regulators | |
2305 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction