Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxM1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074660
Description
FoxM1 Polyclonal specifically detects FoxM1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| FoxM1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| FKHL16trident, forkhead box M1, forkhead box protein M1, Forkhead, drosophila, homolog-like 16, forkhead-like 16, Forkhead-related protein FKHL16, Hepatocyte nuclear factor 3 forkhead homolog 11, HFH11FOXM1B, HFH-11M-phase phosphoprotein 2, HNF-3, HNF-3/fork-head homolog 11, INS-1, MPHOSPH2, MPP-2, MPP2MPM-2 reactive phosphoprotein 2, PIG29, TGT3, Transcription factor Trident, TRIDENT, WIN, Winged-helix factor from INS-1 cells | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. based on sequence homology, this product may cross reacts with Orangutan, Rhesus Monkey, Gorilla, Chinese hamster, and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
| Q08050 | |
| FOXM1 | |
| The immunogen recognized by this antibody maps to a region between residue 713 and 763 of human forkhead box M1 using the numbering given in entry NP_973731.1 (GeneID 2305). | |
| 100 μL | |
| Cancer, Mitotic Regulators | |
| 2305 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction