Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571455
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571455 Supplier Invitrogen™ Supplier No. PA571455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 83%; Guinea Pig: 83%; Human: 100%; Mouse: 91%; Rabbit: 91%; Rat: 91%.

FOXM1 (forkhead box protein M1) is a transcriptional factor regulating the expression of cell cycle genes essential for DNA replication and mitosis. FOXM1 plays a role in the control of cell proliferation and DNA break repairs participating in the DNA damage checkpoint response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXM1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene FOXM1
Gene Accession No. Q08050
Gene Alias AA408308; AW554517; BB238854; cell growth-inhibiting gene 27 protein; FGCP; FKH L16; Fkh16; FKHL 16; FKHL16; folate hydrolase (prostate-specific membrane antigen) 1; Folate hydrolase 1; FOLH; FOLH1; Folylpoly-gamma-glutamate carboxypeptidase; forkhead box M1; forkhead box protein M1; forkhead homolog 16; Forkhead like 16; Forkhead, drosophila, homolog-like 16; forkhead-related protein FKHL16; FOXM 1; FOXM 1c; FOXM1; FOXM1B; GCP2; GCPII; GIG27; glutamate carboxylase II; glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; hepatocyte nuclear factor 3 forkhead homolog 11; HFH 11; HFH11; HFH-11; HFH-11B; HNF 3; HNF3; HNF-3; HNF-3/fork-head homolog 11; INS 1; INS1; INS-1; INS-1 winged helix; Membrane glutamate carboxypeptidase; mGCP; M-phase phosphoprotein 2; MPHOSPH 2; MPHOSPH2; MPM 2; Mpm2; MPM-2 reactive phosphoprotein 2; MPP 2; MPP2; MPP-2; NAALAD1; NAALAdase; NAALADase I; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; PIG 29; PIG29; prostate specific membrane antigen variant F; Prostate-specific membrane antigen; PSM; PSMA; Pteroylpoly-gamma-glutamate carboxypeptidase; TGT3; transcription factor Trident; TRIDENT; WIN; Winged-helix factor from INS-1 cells; winged-helix transcription factor Trident
Gene Symbols FOXM1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human FOXM1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2305
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.