Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527144
Description
Recommended positive controls: FOXM1-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FOXM1 (forkhead box protein M1) is a transcriptional factor regulating the expression of cell cycle genes essential for DNA replication and mitosis. FOXM1 plays a role in the control of cell proliferation and DNA break repairs participating in the DNA damage checkpoint response.
Specifications
FOXM1 | |
Polyclonal | |
Unconjugated | |
FOXM1 | |
AA408308; AW554517; BB238854; cell growth-inhibiting gene 27 protein; FGCP; FKH L16; Fkh16; FKHL 16; FKHL16; folate hydrolase (prostate-specific membrane antigen) 1; Folate hydrolase 1; FOLH; FOLH1; Folylpoly-gamma-glutamate carboxypeptidase; forkhead box M1; forkhead box protein M1; forkhead homolog 16; Forkhead like 16; Forkhead, drosophila, homolog-like 16; forkhead-related protein FKHL16; FOXM 1; FOXM 1c; FOXM1; FOXM1B; GCP2; GCPII; GIG27; glutamate carboxylase II; glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; hepatocyte nuclear factor 3 forkhead homolog 11; HFH 11; HFH11; HFH-11; HFH-11B; HNF 3; HNF3; HNF-3; HNF-3/fork-head homolog 11; INS 1; INS1; INS-1; INS-1 winged helix; Membrane glutamate carboxypeptidase; mGCP; M-phase phosphoprotein 2; MPHOSPH 2; MPHOSPH2; MPM 2; Mpm2; MPM-2 reactive phosphoprotein 2; MPP 2; MPP2; MPP-2; NAALAD1; NAALAdase; NAALADase I; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; PIG 29; PIG29; prostate specific membrane antigen variant F; Prostate-specific membrane antigen; PSM; PSMA; Pteroylpoly-gamma-glutamate carboxypeptidase; TGT3; transcription factor Trident; TRIDENT; WIN; Winged-helix factor from INS-1 cells; winged-helix transcription factor Trident | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2305 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunohistochemistry (Paraffin), Western Blot | |
1.52 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q08050 | |
FOXM1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FOXM1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction