Learn More
FOXM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | FOXM1 |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8520
![]()
|
Proteintech Group Inc
131471AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Product No. 13147-1-AP is rabbit polyclonal antibody raised against C-terminal of human FOXM1. This antibody can recognize three kinds of FOXM1,FOXM1-pp (∽95-110 kDa), FOXM1-B (isoform2,∽83 kDa) and FOXM1A (isoform4, ∽90 kDa)
The protein encoded by this gene is a major non-neuronal microtubule-associated protein. This protein contains a domain similar to the microtubule-binding domains of neuronal microtubule-associated protein (MAP2) and microtubule-associated protein tau (MAPT/TAU). This protein promotes microtubule assembly, and has been shown to counteract destabilization of interphase microtubule catastrophe promotion. Cyclin B was found to interact with this protein, which targets cell division cycle 2 (CDC2) kinase to microtubules. The phosphorylation of this protein affects microtubule properties and cell cycle progression. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| FOXM1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O08696, P97691, Q08050 | |
| 14235, 2305, 58921 | |
| FOXM1 Fusion Protein Ag3729 | |
| Primary | |
| -20°C | |
| FOXM1 |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| FKHL16, forkhead box M1, Forkhead box protein M1, FOXM1, FOXM1B, HFH 11, HFH11, HNF 3, HNF 3/fork head homolog 11, INS 1, M phase phosphoprotein 2, MPHOSPH2, MPP 2, MPP2, PIG29, TGT3, Transcription factor Trident, TRIDENT, WIN | |
| Foxm1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.