Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXN1 Mouse anti-Human, Clone: 1D8B12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | FOXN1 |
---|---|
Clone | 1D8B12 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6768
![]() ![]() |
Proteintech Group Inc
663371IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
FOXN1 is a 648 amino acid protein belonging to the winged helix/forkhead family It contains 1 fork-head DNA-binding domain and is a transcriptional regulator involved in development. FOXN1 is involved in directing lineage choices of multi-potential progenitor epithelial cells and controls important regulating genes of lympho-epithelial interactions. It modulates keratinocyte differentiation in hair follicles and nail apparatus and its defects lead to T-cell immunodeficiency congenital alopecia, nail dystrophy, macroscopic nudity and an inborn dysgenesis of the thymus.Specifications
FOXN1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FKHL20, forkhead box N1, Forkhead box protein N1, FOXN1, RONU, WHN | |
FOXN1 | |
IgG1 | |
Protein G | |
Antibody |
1D8B12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O15353 | |
8456 | |
FOXN1 Fusion Protein Ag18207 | |
Primary | |
-20°C | |
FOXN1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title