Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518191
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518191 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518191 Supplier Invitrogen™ Supplier No. PA518191
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

FOXN1 is a 648 amino acid protein belonging to the winged helix/forkhead family It contains 1 fork-head DNA-binding domain and is a transcriptional regulator involved in development. FOXN1 is involved in directing lineage choices of multi-potential progenitor epithelial cells and controls important regulating genes of lympho-epithelial interactions. It modulates keratinocyte differentiation in hair follicles and nail apparatus and its defects lead to T-cell immunodeficiency congenital alopecia, nail dystrophy, macroscopic nudity and an inborn dysgenesis of the thymus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXN1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene FOXN1
Gene Accession No. O15353
Gene Alias D11Bhm185e; Fkh19; FKHL20; forkhead box N1; forkhead box protein N1; FOXN1; Hepatocyte nuclear factor 3 forkhead homolog 11; Hfh11; HFH-11; HNF-3/forkhead homolog 11; JTK1; nu; nude; Rnu; RONU; Rowett nude; WHN; winged helix nude; winged-helix nude; winged-helix transcription factor nude
Gene Symbols FOXN1
Host Species Goat
Immunogen Synthetic peptide sequence (SVYLSPSSKPVALA) corresponding to the C-terminus amino acids of FOXN1 (aa 635-648).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8456
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.