Learn More
FOXN2 Mouse anti-Human, Clone: OTI7D12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812453

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a forkhead domain binding protein and may function in the transcriptional regulation of the human T-cell leukemia virus long terminal repeat.Specifications
FOXN2 | |
Monoclonal | |
Unconjugated | |
FOXN2 | |
HTLF | |
Mouse | |
Affinity Chromatography | |
RUO | |
3344 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI7D12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P32314 | |
FOXN2 | |
Human recombit protein fragment corresponding to amino acids 1-431 of human FOXN2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.